missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Diaminopyrimidine 98.0+%, TCI America™
$132.35 - $529.40
Chemical Identifiers
| CAS | 2434-56-2 |
|---|---|
| Molecular Formula | C4H6N4 |
| Molecular Weight (g/mol) | 110.12 |
| InChI Key | MISVBCMQSJUHMH-UHFFFAOYSA-N |
| Synonym | 4,6-diaminopyrimidine, 4,6-pyrimidinediamine, zlchem 677, pubchem7074, acmc-1cpb1, ksc206c2f, 4,6-pyrimidinediamine 9ci |
| PubChem CID | 79608 |
| IUPAC Name | pyrimidine-4,6-diamine |
| SMILES | C1=C(N=CN=C1N)N |
Chemical Identifiers
| 2434-56-2 | |
| 110.12 | |
| 4,6-diaminopyrimidine, 4,6-pyrimidinediamine, zlchem 677, pubchem7074, acmc-1cpb1, ksc206c2f, 4,6-pyrimidinediamine 9ci | |
| pyrimidine-4,6-diamine |
| C4H6N4 | |
| MISVBCMQSJUHMH-UHFFFAOYSA-N | |
| 79608 | |
| C1=C(N=CN=C1N)N |
Specifications
| 2434-56-2 | |
| White-Yellow | |
| 1 g | |
| MISVBCMQSJUHMH-UHFFFAOYSA-N | |
| pyrimidine-4,6-diamine | |
| 79608 | |
| ≥98.0% (GC,T) | |
| 4,6-Diaminopyrimidine |
| 271°C | |
| C4H6N4 | |
| 4,6-diaminopyrimidine, 4,6-pyrimidinediamine, zlchem 677, pubchem7074, acmc-1cpb1, ksc206c2f, 4,6-pyrimidinediamine 9ci | |
| C1=C(N=CN=C1N)N | |
| 110.12 | |
| 110.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No