missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diaminopyrimidine 98.0+%, TCI America™

Catalog No. D36481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36481G 1 g
D36485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36481G Supplier TCI America Supplier No. D36481G

Chemical Identifiers

CAS 2434-56-2
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.12
InChI Key MISVBCMQSJUHMH-UHFFFAOYSA-N
Synonym 4,6-diaminopyrimidine, 4,6-pyrimidinediamine, zlchem 677, pubchem7074, acmc-1cpb1, ksc206c2f, 4,6-pyrimidinediamine 9ci
PubChem CID 79608
IUPAC Name pyrimidine-4,6-diamine
SMILES C1=C(N=CN=C1N)N

Specifications

Melting Point 271°C
Color White-Yellow
Quantity 1 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Diaminopyrimidine