missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diaminopyrimidine, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 459800010

 View more versions of this product

Catalog No. AC459800010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4,6-Diaminopyrimidine
Authentic
Glass Bottle
1g
MISVBCMQSJUHMH-UHFFFAOYSA-N
pyrimidine-4,6-diamine
79608
98%
2434-56-2
97.5% min. (HPLC)
C4H6N4
4,6-diaminopyrimidine, 4,6-pyrimidinediamine, zlchem 677, pubchem7074, acmc-1cpb1, ksc206c2f, 4,6-pyrimidinediamine 9ci
C1=C(N=CN=C1N)N
110.1
110.1

Chemical Identifiers

2434-56-2
110.1
4,6-diaminopyrimidine, 4,6-pyrimidinediamine, zlchem 677, pubchem7074, acmc-1cpb1, ksc206c2f, 4,6-pyrimidinediamine 9ci
pyrimidine-4,6-diamine
C4H6N4
MISVBCMQSJUHMH-UHFFFAOYSA-N
79608
C1=C(N=CN=C1N)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only