missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diaminopyrimidine, 98%, Thermo Scientific Chemicals

Catalog No. AC459800050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459800050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC459800050 Supplier Thermo Scientific Chemicals Supplier No. 459800050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2434-56-2 | C4H6N4 | 110.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2434-56-2
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.1
InChI Key MISVBCMQSJUHMH-UHFFFAOYSA-N
Synonym 4,6-diaminopyrimidine, 4,6-pyrimidinediamine, zlchem 677, pubchem7074, acmc-1cpb1, ksc206c2f, 4,6-pyrimidinediamine 9ci
PubChem CID 79608
IUPAC Name pyrimidine-4,6-diamine
SMILES C1=C(N=CN=C1N)N

Specifications

Melting Point 271.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 110.1
Percent Purity 98%
Chemical Name or Material 4,6-Diaminopyrimidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only