missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diamino-2-hydroxy-1,3,5-triazine, tech. 90%

Catalog No. AAL1306403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1306403 1 g
AAL1306406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1306403 Supplier Thermo Scientific Chemicals Supplier No. L1306403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 645-92-1 | C3H5N5O | 127.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 645-92-1
Molecular Formula C3H5N5O
Molecular Weight (g/mol) 127.107
MDL Number MFCD00023186
InChI Key MASBWURJQFFLOO-UHFFFAOYSA-N
Synonym ammeline, 4,6-diamino-1,3,5-triazin-2-ol, ammelin, 1,3,5-triazin-2 1h-one, 4,6-diamino, 2,4-diamino-6-hydroxy-1,3,5-triazine, cyanurodiamide, 2,4-diamino-1,3,5-triazin-6-one, 4,6-diamino-1,3,5-triazin-2 1h-one, unii-lc8g9556q0, s-triazin-2-ol, 4,6-diamino
PubChem CID 12583
ChEBI CHEBI:28646
IUPAC Name 2,6-diamino-1H-1,3,5-triazin-4-one
SMILES C1(=NC(=O)N=C(N1)N)N

Specifications

Quantity 1 g
Beilstein 5570
Formula Weight 127.11
Percent Purity 90%
Grade Technical
Chemical Name or Material 4,6-Diamino-2-hydroxy-1,3,5-triazine
EINECSNumber 211-455-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only