missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4',6'-Diamidino-2-phenylindole dihydrochloride, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 10 mg |
| Solubility Information | (1% in water) Clear |
| IUPAC Name | 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide;dihydrochloride |
| Formula Weight | 350.25 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| EINECSNumber | 249-186-7 |
RUO – Research Use Only