missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4',6-Diamidino-2-phenylindole dihydrochloride, >98%, MP Biomedicals™
$116.25 - $116.25
Chemical Identifiers
| CAS | 28718-90-3 |
|---|---|
| Molecular Formula | C16H17Cl2N5 |
| Molecular Weight (g/mol) | 350.25 |
| MDL Number | MFCD00012681 |
| InChI Key | FPNZBYLXNYPRLR-UHFFFAOYSA-N |
| Synonym | 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836 |
| PubChem CID | 160166 |
| IUPAC Name | dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride |
| SMILES | [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15757405
|
MP Biomedicals Inc
0215757405 |
5 mg |
Each for $116.25
|
|
|||||
Chemical Identifiers
| 28718-90-3 | |
| 350.25 | |
| FPNZBYLXNYPRLR-UHFFFAOYSA-N | |
| 160166 | |
| [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N |
| C16H17Cl2N5 | |
| MFCD00012681 | |
| 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836 | |
| dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride |
Specifications
| 28718-90-3 | |
| >98% | |
| MFCD00012681 | |
| 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836 | |
| [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N | |
| 350.25 | |
| 350.2 | |
| 4', 6-Diamidino-2-phenylindole dihydrochloride |
| Yellow to Orange | |
| C16H17Cl2N5 | |
| 5 mg | |
| FPNZBYLXNYPRLR-UHFFFAOYSA-N | |
| dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride | |
| 160166 | |
| Crystalline Powder |