missing translation for 'onlineSavingsMsg'
Learn More

4,6-Decadiyne 97.0+%, TCI America™

Catalog No. p-7126899
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17261ML 1 mL
D17265ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17261ML Supplier TCI America Supplier No. D17261ML
Only null left

Chemical Identifiers

CAS 16387-71-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00041645
InChI Key LIWZSNTUMSGWTF-UHFFFAOYSA-N
Synonym 4,6-decadiyne, acmc-1c48i
PubChem CID 140067
IUPAC Name deca-4,6-diyne
SMILES CCCC#CC#CCCC

Specifications

Boiling Point 96°C
Quantity 1 mL
UN Number 3295
Formula Weight 134.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,6-Decadiyne
TSCA No
Recommended Storage Refrigerator