missing translation for 'onlineSavingsMsg'
Learn More

4,6-Decadiyne, 97%

Catalog No. p-7035740
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5346703 1 g
AAH5346706 5 g
AAH5346714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5346703 Supplier Thermo Scientific Chemicals Supplier No. H5346703
Only null left

CAS: 16387-71-6 | C10H14 | 134.222 g/mol

6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

Solubility
Immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 16387-71-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00041645
InChI Key LIWZSNTUMSGWTF-UHFFFAOYSA-N
Synonym 4,6-decadiyne, acmc-1c48i
PubChem CID 140067
IUPAC Name deca-4,6-diyne
SMILES CCCC#CC#CCCC

Specifications

Density 0.82
Boiling Point 96°C (15 mmHg)
Linear Formula CH3(CH2)2C≡CC≡C(CH2)2CH3
Quantity 1 g
Solubility Information Immiscible with water.
Formula Weight 134.22
Percent Purity 97%
Chemical Name or Material 4,6-Decadiyne
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only