missing translation for 'onlineSavingsMsg'
Learn More

4-[5-(Trifluoromethyl)pyrid-2-yl]benzonitrile, 97%, Thermo Scientific™

Chemical Identifiers

CAS 892501-99-4
Molecular Formula C13H7F3N2
Molecular Weight (g/mol) 248.21
MDL Number MFCD09064969
InChI Key DFPKSXZRVOQLJC-UHFFFAOYSA-N
Synonym 4-5-trifluoromethyl pyridin-2-yl benzonitrile, 4-5-trifluoromethyl pyrid-2-yl benzonitrile, 4-5-trifluoromethyl-2-pyridinyl benzonitrile, benzonitrile,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl-2-pyridyl benzenecarbonitrile, 4-5-trifluoromethyl pyridin-2-yl benzenecarbonitrile
PubChem CID 24229535
IUPAC Name 4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile
SMILES FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC53016DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC53016DE
View Documents
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

892501-99-4
248.21
DFPKSXZRVOQLJC-UHFFFAOYSA-N
24229535
FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)C#N
C13H7F3N2
MFCD09064969
4-5-trifluoromethyl pyridin-2-yl benzonitrile, 4-5-trifluoromethyl pyrid-2-yl benzonitrile, 4-5-trifluoromethyl-2-pyridinyl benzonitrile, benzonitrile,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl-2-pyridyl benzenecarbonitrile, 4-5-trifluoromethyl pyridin-2-yl benzenecarbonitrile
4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile

Specifications

892501-99-4
100.0
C13H7F3N2
4-5-trifluoromethyl pyridin-2-yl benzonitrile, 4-5-trifluoromethyl pyrid-2-yl benzonitrile, 4-5-trifluoromethyl-2-pyridinyl benzonitrile, benzonitrile,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl-2-pyridyl benzenecarbonitrile, 4-5-trifluoromethyl pyridin-2-yl benzenecarbonitrile
FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)C#N
248.21
248.21
4-[5-(Trifluoromethyl)pyrid-2-yl]benzonitrile
96.5
97%
MFCD09064969
DFPKSXZRVOQLJC-UHFFFAOYSA-N
4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile
24229535
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents