missing translation for 'onlineSavingsMsg'
Learn More

4,5-Methylenedithio-1,3-dithiol-2-one 95.0+%, TCI America™

Catalog No. M1154100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1154100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1154100MG Supplier TCI America Supplier No. M1154100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85720-62-3
Molecular Formula C4H2OS4
Molecular Weight (g/mol) 194.299
MDL Number MFCD00137885
InChI Key VBZVKFOMXXSGFB-UHFFFAOYSA-N
PubChem CID 2060090
IUPAC Name [1,3]dithiolo[4,5-d][1,3]dithiol-5-one
SMILES C1SC2=C(S1)SC(=O)S2

Specifications

Quantity 100 mg
Formula Weight 194.30
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Methylenedithio-1,3-dithiol-2-one