missing translation for 'onlineSavingsMsg'
Learn More

4',5'-Methylenedioxy-2'-nitroacetophenone, 96%

Catalog No. AAL1184103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1184103 1 g
AAL1184106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1184103 Supplier Thermo Scientific Chemicals Supplier No. L1184103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56136-84-6 | C9H7NO5 | 209.157 g/mol

4',5'-Methylenedioxy-2'-nitroacetophenone is used as an intermediate in organic syntheses and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,5′-Methylenedioxy-2′-nitroacetophenone is used as an intermediate in organic syntheses and in chemical research.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 56136-84-6
Molecular Formula C9H7NO5
Molecular Weight (g/mol) 209.157
MDL Number MFCD00053045
InChI Key BQONDGIXVHVIIR-UHFFFAOYSA-N
Synonym 1-6-nitro-1,3-benzodioxol-5-yl ethanone, 4',5'-methylenedioxy-2'-nitroacetophenone, 4,5-methylenedioxy-2-nitroacetophenone, 1-6-nitrobenzo d 1,3 dioxol-5-yl ethanone, 1-6-nitro-1,3-benzodioxol-5-yl ethan-1-one, 1-6-nitro-2h-1,3-benzodioxol-5-yl ethan-1-one, 1-6-nitro-2h-1,3-benzodioxol-5-yl ethanone, 5-acetyl-6-nitro-1,3-benzodioxole, 4.5-methylenedioxy-2-nitroacetophenone, 5-acetyl-6-nitro-2h-benzo d 1,3-dioxolene
PubChem CID 92022
IUPAC Name 1-(6-nitro-1,3-benzodioxol-5-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1[N+](=O)[O-])OCO2

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 199756
Solubility Information Insoluble in water.
Formula Weight 209.16
Percent Purity 98%
Chemical Name or Material 4',5'-Methylenedioxy-2'-nitroacetophenone
EINECSNumber 260-010-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only