missing translation for 'onlineSavingsMsg'
Learn More

4(5)-Iodoimidazole, 94%

Catalog No. p-7048065
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2018403 1 g
AAL2018406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2018403 Supplier Thermo Scientific Chemicals Supplier No. L2018403
Only null left

CAS: 71759-89-2 | C3H3IN2 | 193.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71759-89-2
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.98
MDL Number MFCD01632213
InChI Key BHCMXJKPZOPRNN-UHFFFAOYSA-N
Synonym 4-iodoimidazole, 4-iodo-1h-imidazole, 4 5-iodoimidazole, 4 5-iodo-1 h-imidazole, 4-iodo-1 h-imidazole, 4-iodo-3h-imidazole, 1h-imidazole, 4-iodo, 5-iodoimidazole, 4-iodo-imidazole, sftheqvilimkp@
PubChem CID 606522
IUPAC Name 5-iodo-1H-imidazole
SMILES IC1=CN=CN1

Specifications

Melting Point 135°C to 138°C
Quantity 1 g
Beilstein 1616
Sensitivity Light sensitive
Formula Weight 193.96
Percent Purity 94%
Chemical Name or Material 4(5)-Iodoimidazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only