Learn More
4,5-Imidazoledicarboxylic acid, 97%
CAS: 570-22-9 | C5H4N2O4 | 156.10 g/mol
Supplier: Thermo Scientific Chemicals A1778314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4,5-Imidazoledicarboxylic acid | |
| ∼283°C (decomposition) | |
| C5H4N2O4 | |
| 25 g | |
| 4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid | |
| OC(=O)C1=C(N=CN1)C(O)=O | |
| 156.10 | |
| 156.1 |
| 570-22-9 | |
| 1.749 | |
| MFCD00005200 | |
| 147774 | |
| ZEVWQFWTGHFIDH-UHFFFAOYSA-N | |
| 1H-imidazole-4,5-dicarboxylic acid | |
| 68442 | |
| 97% |
Chemical Identifiers
| 570-22-9 | |
| 156.10 | |
| ZEVWQFWTGHFIDH-UHFFFAOYSA-N | |
| 68442 | |
| OC(=O)C1=C(N=CN1)C(O)=O |
| C5H4N2O4 | |
| MFCD00005200 | |
| 4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid | |
| 1H-imidazole-4,5-dicarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-327-5
RTECSNumber : NI4760000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only