missing translation for 'onlineSavingsMsg'
Learn More

4,5-Imidazoledicarboxylic acid, 97%

Catalog No. AAA1778322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1778322 100 g
AAA1778306 5 g
AAA1778314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1778322 Supplier Thermo Scientific Chemicals Supplier No. A1778322

CAS: 570-22-9 | C5H4N2O4 | 156.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 570-22-9
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00005200
InChI Key ZEVWQFWTGHFIDH-UHFFFAOYSA-N
Synonym 4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid
PubChem CID 68442
IUPAC Name 1H-imidazole-4,5-dicarboxylic acid
SMILES OC(=O)C1=C(N=CN1)C(O)=O

Specifications

Melting Point ∼283°C (decomposition)
Density 1.749
Quantity 100 g
Beilstein 147774
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material 4,5-Imidazoledicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-327-5
RTECSNumber NI4760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only