missing translation for 'onlineSavingsMsg'
Learn More

4(5)-Hydroxymethylimidazole Hydrochloride 98.0+%, TCI America™

Catalog No. H04141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04141G Supplier TCI America Supplier No. H04141G

Chemical Identifiers

CAS 32673-41-9
Molecular Formula C4H7ClN2O
Molecular Weight (g/mol) 134.56
MDL Number MFCD00012697
InChI Key WFNASTYGEKUMIY-UHFFFAOYSA-N
Synonym 4-imidazolemethanol hydrochloride, 4-hydroxymethyl imidazole hydrochloride, 1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethylimidazole hydrochloride, imidazol-4-ylmethanol hydrochloride, 1h-imidazol-4-ylmethanol hydrochloride, 1h-imidazol-5-ylmethanol hydrochloride, 4-hydroxymethyl imidazole hcl, 1h-imidazole-4-ylmethanol hydrochloride, 4 5-hydroxymethylimidazole hydrochloride
PubChem CID 122926
IUPAC Name (1H-imidazol-5-yl)methanol hydrochloride
SMILES Cl.OCC1=CN=CN1

Specifications

Melting Point 108°C
Quantity 1 g
Formula Weight 134.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4(5)-Hydroxymethylimidazole Hydrochloride
TSCA No
Recommended Storage Refrigerator