missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4(5)-Ethylimidazole 98.0+%, TCI America™
$259.11 - $1105.38
Chemical Identifiers
| CAS | 19141-85-6 |
|---|---|
| Molecular Formula | C5H8N2 |
| Molecular Weight (g/mol) | 96.133 |
| InChI Key | NJQHZENQKNIRSY-UHFFFAOYSA-N |
| PubChem CID | 10153736 |
| IUPAC Name | 5-ethyl-1H-imidazole |
| SMILES | CCC1=CN=CN1 |
Chemical Identifiers
| 19141-85-6 | |
| 96.133 | |
| 10153736 | |
| CCC1=CN=CN1 |
| C5H8N2 | |
| NJQHZENQKNIRSY-UHFFFAOYSA-N | |
| 5-ethyl-1H-imidazole |
Specifications
| 19141-85-6 | |
| 140°C | |
| 1 g | |
| CCC1=CN=CN1 | |
| 96.133 | |
| 96.13 | |
| Liquid |
| Yellow | |
| C5H8N2 | |
| NJQHZENQKNIRSY-UHFFFAOYSA-N | |
| 5-ethyl-1H-imidazole | |
| 10153736 | |
| ≥98.0% (GC,T) | |
| 4(5)-Ethylimidazole |
Safety and Handling
TSCA : No