missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-2-furoic acid, 97%, Thermo Scientific™

Catalog No. p-2701040
Change view
Click to view available options
Quantity:
250mg
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB08890CB 250mg Amber Glass Bottle
BTB08890DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB08890CB Supplier Thermo Scientific Chemicals Supplier No. BTB08890CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89639-83-8
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00085031
InChI Key BNVXYXRSJIWWDX-UHFFFAOYSA-N
Synonym 4,5-dimethyl-2-furoic acid, 2-furancarboxylic acid,4,5-dimethyl, 2-furancarboxylic acid, 4,5-dimethyl, 3-methylbenzofurane, acmc-20lon1, 4,5-dimethylfurancarboxylic acid, 4,5-dimethyl-2-furancarboxylic acid, 4,5-dimethyl-furan-2-carboxylic acid
PubChem CID 2774924
IUPAC Name 4,5-dimethylfuran-2-carboxylic acid
SMILES CC1=C(OC(=C1)C(=O)O)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 4, 5-Dimethyl-2-furoic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.