missing translation for 'onlineSavingsMsg'
Learn More

4',5-Dihydroxy-7-methoxyflavone, 97%

Catalog No. p-7046396
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14169MB 25 mg
AAL14169MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14169MB Supplier Thermo Scientific Chemicals Supplier No. L14169MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 437-64-9 | C16H12O5 | 284.267 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 437-64-9
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.267
MDL Number MFCD00017452
InChI Key JPMYFOBNRRGFNO-UHFFFAOYSA-N
Synonym genkwanin, gengkwanin, 4',5-dihydroxy-7-methoxyflavone, 5,4'-dihydroxy-7-methoxyflavone, 7-o-methylapigenin, 7-methylapigenin, apigenin 7-methyl ether, puddumetin, gonkwanin, unii-5k3i5d6b2b
PubChem CID 5281617
ChEBI CHEBI:75718
IUPAC Name 5-hydroxy-2-(4-hydroxyphenyl)-7-methoxychromen-4-one
SMILES COC1=CC(=C2C(=C1)OC(=CC2=O)C3=CC=C(C=C3)O)O

Specifications

Melting Point 288°C to 290°C
Quantity 25 mg
Beilstein 292549
Formula Weight 284.27
Percent Purity 97%
Chemical Name or Material 4',5-Dihydroxy-7-methoxyflavone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only