missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluorosalicylaldehyde, 98%

Catalog No. AAH32680MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32680MD 250 mg
AAH3268003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32680MD Supplier Thermo Scientific Chemicals Supplier No. H32680MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 199287-52-0 | C7H4F2O2 | 158.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 199287-52-0
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.10
MDL Number MFCD08458005
InChI Key HRLWPXGNZQEUHM-UHFFFAOYSA-N
Synonym 2-hydroxy-4,5-difluorobenzaldehyde, 4,5-difluorosalicylaldehyde, 4,5-difluorosalicylaldehdye, benzaldehyde, 4,5-difluoro-2-hydroxy, benzaldehyde, 4,5-difluoro-2-hydroxy-9ci, pubchem11007, 4,5-difluoro-2-hydroxy-benzaldehyde, 4,5-bis fluoranyl-2-oxidanyl-benzaldehyde
PubChem CID 15434553
IUPAC Name 4,5-difluoro-2-hydroxybenzaldehyde
SMILES OC1=CC(F)=C(F)C=C1C=O

Specifications

Melting Point 68°C to 72°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material 4,5-Difluorosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only