missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-nitroaniline 98.0+%, TCI America™

Catalog No. p-7127843
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36445G 5 g
D364425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36445G Supplier TCI America Supplier No. D36445G
Only null left

Chemical Identifiers

CAS 78056-39-0
Molecular Formula C6H4F2N2O2
Molecular Weight (g/mol) 174.11
MDL Number MFCD00010876
InChI Key WDMCABATCGQAKK-UHFFFAOYSA-N
Synonym 3,4-difluoro-6-nitroaniline, benzenamine, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitro-aniline, 4,5-difluoro-2-nitrophenylamine, 2-nitro-4,5-difluoroaniline, 2-amino-4,5-difluoronitrobenzene, 4,5-difluoro-2-nitro-phenylamine, 4,5-difluoro-2-nitrobenzenamine, pubchem2121, acmc-209pdb
PubChem CID 157151
IUPAC Name 4,5-difluoro-2-nitroaniline
SMILES NC1=CC(F)=C(F)C=C1[N+]([O-])=O

Specifications

Melting Point 108°C
Quantity 5 g
Formula Weight 174.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Difluoro-2-nitroaniline