missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-nitroaniline, 98%

Catalog No. p-7037183
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5583003 1 g
AAH5583006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5583003 Supplier Thermo Scientific Chemicals Supplier No. H5583003

CAS: 78056-39-0 | C6H4F2N2O2 | 174.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78056-39-0
Molecular Formula C6H4F2N2O2
Molecular Weight (g/mol) 174.11
MDL Number MFCD00010876
InChI Key WDMCABATCGQAKK-UHFFFAOYSA-N
Synonym 3,4-difluoro-6-nitroaniline, benzenamine, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitro-aniline, 4,5-difluoro-2-nitrophenylamine, 2-nitro-4,5-difluoroaniline, 2-amino-4,5-difluoronitrobenzene, 4,5-difluoro-2-nitro-phenylamine, 4,5-difluoro-2-nitrobenzenamine, pubchem2121, acmc-209pdb
PubChem CID 157151
IUPAC Name 4,5-difluoro-2-nitroaniline
SMILES NC1=CC(F)=C(F)C=C1[N+]([O-])=O

Specifications

Melting Point 107°C to 108°C
Quantity 1 g
Beilstein 2723242
Formula Weight 174.11
Percent Purity 98%
Chemical Name or Material 4,5-Difluoro-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only