missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dicyanoimidazole 98.0+%, TCI America™

Catalog No. p-7127074
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D202625G 25 g
D2026250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D202625G Supplier TCI America Supplier No. D202625G
Only null left

Chemical Identifiers

CAS 1122-28-7
Molecular Formula C5H2N4
Molecular Weight (g/mol) 118.10
MDL Number MFCD00005194
InChI Key XGDRLCRGKUCBQL-UHFFFAOYSA-N
PubChem CID 70729
IUPAC Name 1H-imidazole-4,5-dicarbonitrile
SMILES N#CC1=C(N=CN1)C#N

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Synonym 4,5-dicyanoimidazole, imidazole-4,5-dicarbonitrile, 4,5-imidazoledicarbonitrile, activator, pubchem7602, 4, 5-dicyanoimidazole, moa 7, imidazole-4,5-dinitrile, acmc-1brr7, 4,5-dicyano-1h-imidazole
Formula Weight 118.10
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dicyanoimidazole