missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dicyano-2-phenylimidazole, 97%, Thermo Scientific Chemicals

Catalog No. p-7039527
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5880004 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5880004 Supplier Thermo Scientific Chemicals Supplier No. H5880004
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50847-06-8
Molecular Formula C11H6N4
Molecular Weight (g/mol) 194.20
MDL Number MFCD05863325
InChI Key PGSHQANDAMACEF-UHFFFAOYSA-N
Synonym 2-phenyl-4,5-dicyanoimidazole, 2-phenylimidazole-4,5-dicarbonitrile, phenylimidazoledicarbonitrile, 2-phenyl-1h-imidazol-4,5-dikohlenitrile, 1h-imidazole-4,5-dicarbonitrile,2-phenyl, 1h-imidazole-4,5-dicarbonitrile, 2-phenyl
PubChem CID 820939
IUPAC Name 2-phenyl-1H-imidazole-4,5-dicarbonitrile
SMILES N#CC1=C(N=C(N1)C1=CC=CC=C1)C#N

Specifications

Quantity 2 g
UN Number UN3439
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 4,5-Dicyano-2-phenylimidazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only