missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloroimidazole, 98%

Catalog No. AAL0626206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0626206 5 g
AAL0626214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0626206 Supplier Thermo Scientific Chemicals Supplier No. L0626206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15965-30-7 | C3H2Cl2N2 | 136.96 g/mol

4,5-Dichloroimidazole has been used in the synthesis of: phosphinite complexes, 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dichloroimidazole has been used in the synthesis of: phosphinite complexes, 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, strong acids.

Chemical Identifiers

CAS 15965-30-7
Molecular Formula C3H2Cl2N2
Molecular Weight (g/mol) 136.96
MDL Number MFCD00005195
InChI Key QAJJXHRQPLATMK-UHFFFAOYSA-N
Synonym 4,5-dichloroimidazole, 1h-imidazole, 4,5-dichloro, 4,5-dichlorimidazole, pubchem12880, acmc-1bqkp, 3,4-dichloro-2h-pyrazole, ksc530c6j, 1h-imidazole,4,5-dichloro, qajjxhrqplatmk-uhfffaoysa
PubChem CID 140018
IUPAC Name 4,5-dichloro-1H-imidazole
SMILES ClC1=C(Cl)N=CN1

Specifications

Melting Point 179°C to 182°C
Quantity 5 g
Beilstein 606155
Solubility Information Insoluble in water.
Formula Weight 136.97
Percent Purity 98%
Chemical Name or Material 4,5-Dichloroimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only