missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-1,2-phenylenediamine 98.0+%, TCI America™

Catalog No. p-7127220
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23075G 5 g
D230725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23075G Supplier TCI America Supplier No. D23075G

Chemical Identifiers

CAS 5348-42-5
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.03
MDL Number MFCD00007723
InChI Key IWFHBRFJOHTIPU-UHFFFAOYSA-N
Synonym 4,5-dichloro-o-phenylenediamine, 4,5-dichloro-1,2-benzenediamine, 1,2-diamino-4,5-dichlorobenzene, 4,5-dichloro-1,2-phenylenediamine, 1,2-benzenediamine, 4,5-dichloro, 4,5-dichloro-o-phenylendiamine, 1,2-dichloro-4,5-diaminobenzene, 2-amino-4,5-dichloroaniline, 4,5-dichloro-ortho-phenylenediamine, 4,5-dichlorophenylene-1,2-diamine
PubChem CID 79297
IUPAC Name 4,5-dichlorobenzene-1,2-diamine
SMILES NC1=CC(Cl)=C(Cl)C=C1N

Specifications

Melting Point 163°C
Quantity 5 g
Formula Weight 177.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dichloro-1,2-phenylenediamine