missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromothiophene-2-sulfonyl chloride, 97%

CAS: 81606-31-7 | C4HBr2ClO2S2 | 340.43 g/mol

Supplier:  Thermo Scientific Chemicals A1338414

 View more versions of this product

Catalog No. AAA1338414


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4,5-Dibromothiophene-2-sulfonyl chloride
75°C to 79°C
MFCD00041439
UN3261
Moisture sensitive
WJYGHWXWQSCONR-UHFFFAOYSA-N
4,5-dibromothiophene-2-sulfonyl chloride
522755
97%
81606-31-7
C4HBr2ClO2S2
25 g
162960
4,5-dibromothiophene-2-sulphonyl chloride, 2,3-dibromothiophene-5-sulfonyl chloride, 4,5-dibromo-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dibromo, 4,5-dibromothiophene-2-sulfonic acid chloride, acmc-20ams9, buttpark 9350-57, 4,5-dibromo 2-thienyl chlorosulfone, 2,3-dibromothiophene-5-sulfonylchloride, 4,5-dibromothiophene-2-sulfonylchloride
ClS(=O)(=O)C1=CC(Br)=C(Br)S1
340.43
340.45

Chemical Identifiers

81606-31-7
340.43
WJYGHWXWQSCONR-UHFFFAOYSA-N
522755
ClS(=O)(=O)C1=CC(Br)=C(Br)S1
C4HBr2ClO2S2
MFCD00041439
4,5-dibromothiophene-2-sulphonyl chloride, 2,3-dibromothiophene-5-sulfonyl chloride, 4,5-dibromo-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dibromo, 4,5-dibromothiophene-2-sulfonic acid chloride, acmc-20ams9, buttpark 9350-57, 4,5-dibromo 2-thienyl chlorosulfone, 2,3-dibromothiophene-5-sulfonylchloride, 4,5-dibromothiophene-2-sulfonylchloride
4,5-dibromothiophene-2-sulfonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only