missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB07379DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. BTB07379DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

14305-08-9
329.979
NQJXEMRMTOSSBQ-UHFFFAOYSA-N
203396
C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Br)Br
C10H6Br2N2O
MFCD00052907
4,5-dibromo-2-phenylpyridazin-3 2h-one, 4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 4,5-dibromo-2-phenyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dibromo-2-phenyl, 4,5-dibromo-2-phenyl-3-pyridazinone, 4,5-dibromo-2-phenyl-pyridazin-3-one, 2-phenyl-4,5-dibromo-3 2h-pyridazinone, 3 2h-pyridazinone,4,5-dibromo-2-phenyl, 4,5-bis bromanyl-2-phenyl-pyridazin-3-one, 4,5-dibromo-2-phenyl-2-hydropyridazin-3-one
4,5-dibromo-2-phenylpyridazin-3-one

Specifications

4, 5-Dibromo-2-phenyl-2, 3-dihydropyridazin-3-one
96.5
97%
C10H6Br2N2O
MFCD00052907
NQJXEMRMTOSSBQ-UHFFFAOYSA-N
4,5-dibromo-2-phenylpyridazin-3-one
203396
97%
14305-08-9
100.0
Amber Glass Bottle
1g
4,5-dibromo-2-phenylpyridazin-3 2h-one, 4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 4,5-dibromo-2-phenyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dibromo-2-phenyl, 4,5-dibromo-2-phenyl-3-pyridazinone, 4,5-dibromo-2-phenyl-pyridazin-3-one, 2-phenyl-4,5-dibromo-3 2h-pyridazinone, 3 2h-pyridazinone,4,5-dibromo-2-phenyl, 4,5-bis bromanyl-2-phenyl-pyridazin-3-one, 4,5-dibromo-2-phenyl-2-hydropyridazin-3-one
C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Br)Br
329.979
329.98
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions