missing translation for 'onlineSavingsMsg'
Learn More

4,5-Diazafluoren-9-one 98.0+%, TCI America™

Catalog No. D4255200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4255200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4255200MG Supplier TCI America Supplier No. D4255200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50890-67-0
Molecular Formula C11H6N2O
Molecular Weight (g/mol) 182.182
MDL Number MFCD00046892
InChI Key PFMTUGNLBQSHQC-UHFFFAOYSA-N
Synonym 4,5-diazafluoren-9-one, 5h-cyclopenta 1,2-b:5,4-b' dipyridin-5-one, 5h-cyclopenta 2,1-b:3,4-b' dipyridin-5-one, 5h-pyrido 3',2':4,5 cyclopenta 1,2-b pyridin-5-one, pyridino 3',2'-1,2 cyclopenta 3,4-b pyridin-5-one, dafone, dafo, 4,5-diazafluorenone-0, 4,5-diaza-9h-fluoren-9-one
PubChem CID 342157
SMILES C1=CC2=C(C3=C(C2=O)C=CC=N3)N=C1

Specifications

Melting Point 216°C
Quantity 200 mg
Formula Weight 182.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Diazafluoren-9-one