missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Bis(methylthio)-1,3-dithiole-2-thione 98.0+%, TCI America™
Supplier: TCI America B17781G
Specifications
| 4,5-Bis(methylthio)-1,3-dithiole-2-thione | |
| 102°C | |
| C5H6S5 | |
| 1 g | |
| CSC1=C(SC(=S)S1)SC | |
| 226.403 | |
| 226.40 | |
| Crystalline Powder |
| 49638-64-4 | |
| Yellow | |
| MFCD00137888 | |
| HOFVXSUZSDYZSA-UHFFFAOYSA-N | |
| 4,5-bis(methylsulfanyl)-1,3-dithiole-2-thione | |
| 362959 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 49638-64-4 | |
| 226.403 | |
| HOFVXSUZSDYZSA-UHFFFAOYSA-N | |
| 4,5-bis(methylsulfanyl)-1,3-dithiole-2-thione |
| C5H6S5 | |
| MFCD00137888 | |
| 362959 | |
| CSC1=C(SC(=S)S1)SC |
Safety and Handling
TSCA : No