missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5,9,10-Tetrahydropyrene 98.0+%, TCI America™
$91.33 - $885.43
Chemical Identifiers
| CAS | 781-17-9 |
|---|---|
| Molecular Formula | C16H14 |
| Molecular Weight (g/mol) | 206.288 |
| MDL Number | MFCD00267195 |
| InChI Key | XDFUNRTWHPWCKO-UHFFFAOYSA-N |
| PubChem CID | 69906 |
| IUPAC Name | 4,5,9,10-tetrahydropyrene |
| SMILES | C1CC2=C3C(=CC=C2)CCC4=CC=CC1=C43 |
Chemical Identifiers
| 781-17-9 | |
| 206.288 | |
| XDFUNRTWHPWCKO-UHFFFAOYSA-N | |
| 4,5,9,10-tetrahydropyrene |
| C16H14 | |
| MFCD00267195 | |
| 69906 | |
| C1CC2=C3C(=CC=C2)CCC4=CC=CC1=C43 |
Specifications
| 781-17-9 | |
| C16H14 | |
| 100 mg | |
| C1CC2=C3C(=CC=C2)CCC4=CC=CC1=C43 | |
| 206.288 | |
| 206.29 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00267195 | |
| XDFUNRTWHPWCKO-UHFFFAOYSA-N | |
| 4,5,9,10-tetrahydropyrene | |
| 69906 | |
| ≥98.0% (GC) | |
| 4,5,9,10-Tetrahydropyrene |
Safety and Handling
TSCA : No