missing translation for 'onlineSavingsMsg'
Learn More

4,5,9,10-Tetrahydropyrene 98.0+%, TCI America™

Catalog No. T2264100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2264100MG 100 mg
T22641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2264100MG Supplier TCI America Supplier No. T2264100MG
Only null left

Chemical Identifiers

CAS 781-17-9
Molecular Formula C16H14
Molecular Weight (g/mol) 206.288
MDL Number MFCD00267195
InChI Key XDFUNRTWHPWCKO-UHFFFAOYSA-N
PubChem CID 69906
IUPAC Name 4,5,9,10-tetrahydropyrene
SMILES C1CC2=C3C(=CC=C2)CCC4=CC=CC1=C43

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 206.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5,9,10-Tetrahydropyrene