missing translation for 'onlineSavingsMsg'
Learn More

4,5,9,10-Tetrahydropyrene (purified by sublimation) 98.0+%, TCI America™

Catalog No. T2655100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2655100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2655100MG Supplier TCI America Supplier No. T2655100MG
Only null left

Chemical Identifiers

CAS 781-17-9
Molecular Formula C16H14
Molecular Weight (g/mol) 206.288
MDL Number MFCD00267195
InChI Key XDFUNRTWHPWCKO-UHFFFAOYSA-N
PubChem CID 69906
IUPAC Name 4,5,9,10-tetrahydropyrene
SMILES C1CC2=C3C(=CC=C2)CCC4=CC=CC1=C43

Specifications

Melting Point 140°C
Color White-Yellow
Boiling Point 383°C
Quantity 100 mg
Formula Weight 206.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5,9,10-Tetrahydropyrene (purified by sublimation)