Learn More
4,5',8-Trimethylpsoralen, 99%
CAS: 3902-71-4 | C14H12O3 | 228.25 g/mol
Supplier: Thermo Scientific Chemicals J6322606
Description
4,5',8-Trimethylpsoralen is used for treatment of psoriasis, and as a photochemical crosslinker of DNA used as a probe for nucleic acid structure, used to crosslink DNA onto mica surfaces, mutagenesis of N2 worms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,5′,8-Trimethylpsoralen is used for treatment of psoriasis, and as a photochemical crosslinker of DNA used as a probe for nucleic acid structure, used to crosslink DNA onto mica surfaces, mutagenesis of N2 worms.
Solubility
Soluble in chloroform
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications
| 4,5',8-Trimethylpsoralen | |
| 229°C to 231°C | |
| C14H12O3 | |
| 5 g | |
| 221723 | |
| trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen | |
| FMHHVULEAZTJMA-UHFFFAOYSA-N | |
| 2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one | |
| 5585 | |
| 228.25 | |
| Powder |
| 3902-71-4 | |
| White | |
| MFCD00005010 | |
| UN1759 | |
| 14,9735 | |
| Soluble in chloroform | |
| CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1 | |
| 228.25 | |
| CHEBI:28329 | |
| 99% |
Chemical Identifiers
| 3902-71-4 | |
| 228.25 | |
| FMHHVULEAZTJMA-UHFFFAOYSA-N | |
| 5585 | |
| 2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one |
| C14H12O3 | |
| MFCD00005010 | |
| trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen | |
| CHEBI:28329 | |
| CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1 |
Safety and Handling
GHS H Statement
H314-H318-H351
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H351
DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 223-459-0
RTECSNumber : LV1576000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only