missing translation for 'onlineSavingsMsg'
Learn More

4',5,7-Trihydroxyflavanone, 97%

Catalog No. AAL0983404
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0983404 2 g
AAL0983409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0983404 Supplier Thermo Scientific Chemicals Supplier No. L0983404
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 67604-48-2 | C15H12O5 | 272.256 g/mol

(±)-Naringenin is an antioxidant flavonoid. It has anti-inflammatory and antitumor properties. (±)-Naringenin induces apoptosis. It stimulates DNA repair following oxidative damage. (±)-Naringenin inhibits the activity of PI 3-kinase (PI(3)K).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-Naringenin is an antioxidant flavonoid. It has anti-inflammatory and antitumor properties. (±)-Naringenin induces apoptosis. It stimulates DNA repair following oxidative damage. (±)-Naringenin inhibits the activity of PI 3-kinase (PI(3)K).

Solubility
Soluble in ethanol (50 mg/ml), DMSO, methanol, and ammonium hydroxide (50 mg/ml). Insoluble in water (almost)

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 67604-48-2
Molecular Formula C15H12O5
Molecular Weight (g/mol) 272.256
MDL Number MFCD00006844
InChI Key FTVWIRXFELQLPI-UHFFFAOYSA-N
Synonym naringenin, 5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, naringenine, 4',5,7-trihydroxyflavanone, +/--naringenin, naringetol, salipurpol, --naringenin, narigenin, salipurol
PubChem CID 932
ChEBI CHEBI:50202
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O

Specifications

Melting Point 248°C to 251°C
Quantity 2 g
Merck Index 14,6424
Solubility Information Soluble in ethanol (50mg/ml),DMSO,methanol,and ammonium hydroxide (50mg/ml). Insoluble in water (almost)
Formula Weight 272.26
Percent Purity 97%
Chemical Name or Material 4',5,7-Trihydroxyflavanone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 266-769-1
RTECSNumber DJ2981530
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only