missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carboxylic acid, 97%

Catalog No. p-4228909
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432670010 1 g Glass bottle
AC432670050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432670010 Supplier Thermo Scientific Chemicals Supplier No. 432670010

CAS: 307307-97-7 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 307307-97-7
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD09056813
InChI Key VOAKNFVZEGNOKV-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carboxylic acid, 4h,5h,6h,7h-pyrazolo 1,5-a pyridine-3-carboxylic acid, pyrazolo 1,5-a pyridine-3-carboxylic acid, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-pyrazolo 1,5-a pyridine-3-carboxylic acid, pubchem23924, 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carbox, 4?5?6?7-tetrahydropyrazolo 1?5-a pyridine-3-carboxylic acid, pyrazolo 1,5-a pyridine-3-carboxylicacid, 4,5,6,7-tetrahydro
PubChem CID 21941376
IUPAC Name 4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-3-carboxylic acid
SMILES C1CCN2C(=C(C=N2)C(=O)O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 215°C to 223°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 166.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only