missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid, 97%

Catalog No. AAH3306203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3306203 1 g
AAH3306206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3306203 Supplier Thermo Scientific Chemicals Supplier No. H3306203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 40133-07-1 | C9H10O2S | 182.237 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 40133-07-1
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00464469
InChI Key ROLXOQXKNDKXTA-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro, acmc-1asbb, enamine_001826, 2-carboxy-4,5,6,7-tetrahydro-1-benzothiophene, 2-carboxy-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylicacid, 4,5,6,7-tetrahydrobenzo b-thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic
PubChem CID 2063443
IUPAC Name 4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylic acid
SMILES C1CCC2=C(C1)C=C(S2)C(=O)O

Specifications

Melting Point 191°C to 193°C
Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 182.24
Percent Purity 97%
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only