missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride, 95%

Catalog No. p-7056018
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64755MD 250 mg
AAH6475503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64755MD Supplier Thermo Scientific Chemicals Supplier No. H64755MD
Only null left

CAS: 879668-17-4 | C6H10ClN3 | 159.617 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 879668-17-4
Molecular Formula C6H10ClN3
Molecular Weight (g/mol) 159.617
MDL Number MFCD11846200
InChI Key CHLFJSVTUQAGSC-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl, 4,5,6,7-tetrahydro-1h-imidazo 4,5-c pyridine hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-5-azabenzimidazole hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloridyl, 1h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-1h-imidazo 3,4-c pyridine hydrochloride, 4,5,6,7-tetrahydro-imidazo-4,5-c-pyridine hydrochloride
PubChem CID 21330723
IUPAC Name 4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine;hydrochloride
SMILES C1CNCC2=C1N=CN2.Cl

Specifications

Quantity 250 mg
Beilstein 15312806
Formula Weight 159.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only