missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5,6,7-Tetrabromobenzotriazole 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 264°C |
| Color | White-Yellow |
| Quantity | 50 mg |
| Formula Weight | 434.71 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,5,6,7-Tetrabromobenzotriazole |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |