missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[4-(Trifluoromethoxy)phenoxy]piperidine 97.0+%, TCI America™
$111.66 - $377.33
Chemical Identifiers
| CAS | 287952-67-4 |
|---|---|
| Molecular Formula | C12H14F3NO2 |
| Molecular Weight (g/mol) | 261.24 |
| MDL Number | MFCD06656166 |
| InChI Key | RPQOTFPZKNHYFB-UHFFFAOYSA-N |
| Synonym | 4-4-trifluoromethoxy phenoxy piperidine, 4-4-trifluoromethoxyphenoxy piperidine, piperidine, 4-4-trifluoromethoxy phenoxy, 4-4-trifluoromethoxy-phenoxy-piperidine, acmc-1cpm9, 4-4-trifluoromethoxy-phenoxy piperidine, 4-4-trifluoromethoxy phenoxy-piperidine |
| PubChem CID | 16115439 |
| IUPAC Name | 4-[4-(trifluoromethoxy)phenoxy]piperidine |
| SMILES | FC(F)(F)OC1=CC=C(OC2CCNCC2)C=C1 |
Chemical Identifiers
| 287952-67-4 | |
| 261.24 | |
| RPQOTFPZKNHYFB-UHFFFAOYSA-N | |
| 16115439 | |
| FC(F)(F)OC1=CC=C(OC2CCNCC2)C=C1 |
| C12H14F3NO2 | |
| MFCD06656166 | |
| 4-4-trifluoromethoxy phenoxy piperidine, 4-4-trifluoromethoxyphenoxy piperidine, piperidine, 4-4-trifluoromethoxy phenoxy, 4-4-trifluoromethoxy-phenoxy-piperidine, acmc-1cpm9, 4-4-trifluoromethoxy-phenoxy piperidine, 4-4-trifluoromethoxy phenoxy-piperidine | |
| 4-[4-(trifluoromethoxy)phenoxy]piperidine |
Specifications
| 287952-67-4 | |
| White-Yellow | |
| MFCD06656166 | |
| 4-4-trifluoromethoxy phenoxy piperidine, 4-4-trifluoromethoxyphenoxy piperidine, piperidine, 4-4-trifluoromethoxy phenoxy, 4-4-trifluoromethoxy-phenoxy-piperidine, acmc-1cpm9, 4-4-trifluoromethoxy-phenoxy piperidine, 4-4-trifluoromethoxy phenoxy-piperidine | |
| FC(F)(F)OC1=CC=C(OC2CCNCC2)C=C1 | |
| 261.24 | |
| 261.24 | |
| Crystalline Powder |
| 76°C | |
| C12H14F3NO2 | |
| 1 g | |
| RPQOTFPZKNHYFB-UHFFFAOYSA-N | |
| 4-[4-(trifluoromethoxy)phenoxy]piperidine | |
| 16115439 | |
| ≥97.0% (GC,T) | |
| 4-[4-(Trifluoromethoxy)phenoxy]piperidine |
Safety and Handling
TSCA : No