missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(Trifluoromethoxy)phenoxy]piperidine 97.0+%, TCI America™

Catalog No. T33495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33495G 5 g
T33491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33495G Supplier TCI America Supplier No. T33495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 287952-67-4
Molecular Formula C12H14F3NO2
Molecular Weight (g/mol) 261.24
MDL Number MFCD06656166
InChI Key RPQOTFPZKNHYFB-UHFFFAOYSA-N
Synonym 4-4-trifluoromethoxy phenoxy piperidine, 4-4-trifluoromethoxyphenoxy piperidine, piperidine, 4-4-trifluoromethoxy phenoxy, 4-4-trifluoromethoxy-phenoxy-piperidine, acmc-1cpm9, 4-4-trifluoromethoxy-phenoxy piperidine, 4-4-trifluoromethoxy phenoxy-piperidine
PubChem CID 16115439
IUPAC Name 4-[4-(trifluoromethoxy)phenoxy]piperidine
SMILES FC(F)(F)OC1=CC=C(OC2CCNCC2)C=C1

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 5 g
Formula Weight 261.24
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[4-(Trifluoromethoxy)phenoxy]piperidine