missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridylmethyl)aniline, 97%, Thermo Scientific™

$135.34 - $135.34

Chemical Identifiers

CAS 27692-74-6
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00053060
InChI Key WZXYYQHVDJMIFF-UHFFFAOYSA-N
Synonym 4-4-pyridylmethyl aniline, 4-pyridin-4-ylmethyl aniline, 4-pyridin-4-ylmethyl-phenylamine, benzenamine, 4-4-pyridinylmethyl, 4-4-pyridinylmethyl aniline, 4-pyridin-4-yl methyl aniline, 4-pyridin-4-ylmethyl phenyl amine, 4-4-pyridylmethyl phenylamine, 4-p-aminobenzylpyridine, enamine_005904
PubChem CID 307158
IUPAC Name 4-(pyridin-4-ylmethyl)aniline
SMILES C1=CC(=CC=C1CC2=CC=NC=C2)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB01214DA
SDS View Documents
Thermo Scientific Chemicals
BTB01214DA
1 g Amber Glass Bottle
Each for $135.34
Only null left
 
Description

Description

Specifications

Chemical Identifiers

27692-74-6
184.242
WZXYYQHVDJMIFF-UHFFFAOYSA-N
307158
C1=CC(=CC=C1CC2=CC=NC=C2)N
C12H12N2
MFCD00053060
4-4-pyridylmethyl aniline, 4-pyridin-4-ylmethyl aniline, 4-pyridin-4-ylmethyl-phenylamine, benzenamine, 4-4-pyridinylmethyl, 4-4-pyridinylmethyl aniline, 4-pyridin-4-yl methyl aniline, 4-pyridin-4-ylmethyl phenyl amine, 4-4-pyridylmethyl phenylamine, 4-p-aminobenzylpyridine, enamine_005904
4-(pyridin-4-ylmethyl)aniline

Specifications

27692-74-6
100.0
Amber Glass Bottle
MFCD00053060
4-4-pyridylmethyl aniline, 4-pyridin-4-ylmethyl aniline, 4-pyridin-4-ylmethyl-phenylamine, benzenamine, 4-4-pyridinylmethyl, 4-4-pyridinylmethyl aniline, 4-pyridin-4-yl methyl aniline, 4-pyridin-4-ylmethyl phenyl amine, 4-4-pyridylmethyl phenylamine, 4-p-aminobenzylpyridine, enamine_005904
C1=CC(=CC=C1CC2=CC=NC=C2)N
184.242
184.24
Technical
96.5
97%
C12H12N2
1 g
WZXYYQHVDJMIFF-UHFFFAOYSA-N
4-(pyridin-4-ylmethyl)aniline
307158
97%
4-(4-Pyridylmethyl)aniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'