missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridylmethyl)aniline, 97%, Thermo Scientific™

Catalog No. BTB01214DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB01214DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB01214DA Supplier Thermo Scientific Chemicals Supplier No. BTB01214DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27692-74-6
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00053060
InChI Key WZXYYQHVDJMIFF-UHFFFAOYSA-N
Synonym 4-4-pyridylmethyl aniline, 4-pyridin-4-ylmethyl aniline, 4-pyridin-4-ylmethyl-phenylamine, benzenamine, 4-4-pyridinylmethyl, 4-4-pyridinylmethyl aniline, 4-pyridin-4-yl methyl aniline, 4-pyridin-4-ylmethyl phenyl amine, 4-4-pyridylmethyl phenylamine, 4-p-aminobenzylpyridine, enamine_005904
PubChem CID 307158
IUPAC Name 4-(pyridin-4-ylmethyl)aniline
SMILES C1=CC(=CC=C1CC2=CC=NC=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.24
Percent Purity 97%
Grade Technical
Chemical Name or Material 4-(4-Pyridylmethyl)aniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.