missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridyl)morpholine 98.0+%, TCI America™

Catalog No. P16465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16465G 5 g
P164625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16465G Supplier TCI America Supplier No. P16465G
Only null left

Chemical Identifiers

CAS 2767-91-1
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
InChI Key QJWQYVJVCXMTJP-UHFFFAOYSA-N
Synonym 4-morpholinopyridine, 4-pyridin-4-yl morpholine, 4-4-pyridyl morpholine, morpholine, 4-4-pyridinyl, acmc-1cmri, 4-4-pyridinyl morpholine, 4-4-morpholinyl pyridine
PubChem CID 4145413
IUPAC Name 4-pyridin-4-ylmorpholine
SMILES C1COCCN1C2=CC=NC=C2

Specifications

Melting Point 107°C
Quantity 5 g
Formula Weight 164.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Pyridyl)morpholine