missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Pyridyl)benzaldehyde 98.0+%, TCI America™
Supplier: TCI America P19831G
Specifications
| 4-(4-Pyridyl)benzaldehyde | |
| 89°C | |
| C12H9NO | |
| 1 g | |
| MBJXDIYHLGBQOT-UHFFFAOYSA-N | |
| 4-pyridin-4-ylbenzaldehyde | |
| 639673 | |
| ≥98.0% (GC) |
| 99163-12-9 | |
| White-Yellow | |
| MFCD02684110 | |
| 4-pyridin-4-yl-benzaldehyde, 4-pyridin-4-yl benzaldehyde, 4-4-pyridyl benzaldehyde, 4-4-pyridinyl benzaldehyde, 4-4-formylphenyl pyridine, akos bar-0607, benzaldehyde, 4-4-pyridinyl, acmc-209sbt | |
| C1=CC(=CC=C1C=O)C2=CC=NC=C2 | |
| 183.21 | |
| 183.21 | |
| Crystalline Powder |
Chemical Identifiers
| 99163-12-9 | |
| 183.21 | |
| MBJXDIYHLGBQOT-UHFFFAOYSA-N | |
| 639673 | |
| C1=CC(=CC=C1C=O)C2=CC=NC=C2 |
| C12H9NO | |
| MFCD02684110 | |
| 4-pyridin-4-yl-benzaldehyde, 4-pyridin-4-yl benzaldehyde, 4-4-pyridyl benzaldehyde, 4-4-pyridinyl benzaldehyde, 4-4-formylphenyl pyridine, akos bar-0607, benzaldehyde, 4-4-pyridinyl, acmc-209sbt | |
| 4-pyridin-4-ylbenzaldehyde |
Safety and Handling
TSCA : No