missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridyl)benzaldehyde 98.0+%, TCI America™

Catalog No. P1983200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1983200MG 200 mg
P19831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1983200MG Supplier TCI America Supplier No. P1983200MG

Chemical Identifiers

CAS 99163-12-9
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD02684110
InChI Key MBJXDIYHLGBQOT-UHFFFAOYSA-N
Synonym 4-pyridin-4-yl-benzaldehyde, 4-pyridin-4-yl benzaldehyde, 4-4-pyridyl benzaldehyde, 4-4-pyridinyl benzaldehyde, 4-4-formylphenyl pyridine, akos bar-0607, benzaldehyde, 4-4-pyridinyl, acmc-209sbt
PubChem CID 639673
IUPAC Name 4-pyridin-4-ylbenzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=CC=NC=C2

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 200 mg
Formula Weight 183.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Pyridyl)benzaldehyde