missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Pyridyl)-2,2':6',2″-terpyridine 97.0+%, TCI America™

Catalog No. P2202200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2202200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2202200MG Supplier TCI America Supplier No. P2202200MG

Chemical Identifiers

CAS 112881-51-3
Molecular Formula C20H14N4
Molecular Weight (g/mol) 310.36
MDL Number MFCD00932037
InChI Key DPPKPCLKZOLTMB-UHFFFAOYSA-N
PubChem CID 11438308
IUPAC Name 2,6-dipyridin-2-yl-4-pyridin-4-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=NC=C4

Specifications

Melting Point 223°C
Color Yellow
Quantity 200 mg
Formula Weight 310.36
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(4-Pyridyl)-2,2′:6′,2′′-terpyridine