missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Piperidyl)pyridine 98.0+%, TCI America™

Catalog No. P20121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20121G 1 g
P20125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20121G Supplier TCI America Supplier No. P20121G
Only null left

Chemical Identifiers

CAS 581-45-3
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD05863877
InChI Key RGBWBVGQZFJTEO-UHFFFAOYSA-N
Synonym 4-(4-Pyridyl)piperidine
PubChem CID 1139597
IUPAC Name 4-piperidin-4-ylpyridine
SMILES C1CNCCC1C2=CC=NC=C2

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 292°C
Quantity 1 g
Formula Weight 162.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Piperidyl)pyridine