missing translation for 'onlineSavingsMsg'
Learn More

4,4-Pentamethylene-2-pyrrolidone 98.0+%, TCI America™

Catalog No. p-7135214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16955G 5 g
P169525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16955G Supplier TCI America Supplier No. P16955G
Only null left

Chemical Identifiers

CAS 64744-50-9
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.23
MDL Number MFCD00177938
InChI Key JAWPQJDOQPSNIQ-UHFFFAOYSA-N
Synonym 3-Azaspiro[4.5]decan-2-one
PubChem CID 47457
IUPAC Name 2-azaspiro[4.5]decan-3-one
SMILES O=C1CC2(CN1)CCCCC2

Specifications

Melting Point 91°C
Color White-Yellow
Boiling Point 181°C
Quantity 5 g
Formula Weight 153.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Pentamethylene-2-pyrrolidone
RTECSNumber CM0120000
TSCA No