missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenylazo)resorcinol, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 151510250

 View more versions of this product

Catalog No. AC151510250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-(4-Nitrophenylazo)resorcinol
74-39-5
88% min. (HPLC)
C12H9N3O4
25g
15, 5759
Solubility in water: practically insoluble. Other solubilities: soluble in dilute aquous naoh
OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O
259.22
259.21
dye content ca 90%
Authentic
Glass bottle
MFCD00007310
16, 181
p-nitrophenylazoresorcinol, p-diazoviolet, p-nitrobenzeneazoresorcinol, magneson i, magnezon i, 4-p-nitrophenylazo resorcinol, p-diazviolet, 2,4-dihydroxy-4'-nitroazobenzene, magneson, 4-4-nitrophenylazo resorcinol
BIFFKKLAAIGZBV-UHFFFAOYSA-N
(4E)-3-hydroxy-4-[(4-nitrophenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
5717413
90%

Chemical Identifiers

74-39-5
259.22
BIFFKKLAAIGZBV-UHFFFAOYSA-N
5717413
OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O
C12H9N3O4
MFCD00007310
p-nitrophenylazoresorcinol, p-diazoviolet, p-nitrobenzeneazoresorcinol, magneson i, magnezon i, 4-p-nitrophenylazo resorcinol, p-diazviolet, 2,4-dihydroxy-4'-nitroazobenzene, magneson, 4-4-nitrophenylazo resorcinol
(4E)-3-hydroxy-4-[(4-nitrophenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 200-808-5

RTECSNumber : VH2810000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only